Assay Details
| Gene Symbol: Kcnj5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010605.4 | NP_034735.3 | 1 - 2 | 137 | 64 | |
| GenBank mRNA | AK052891.1 | - | 1 - 2 | 138 | 64 | |
| AK137600.1 | - | 1 - 2 | 251 | 64 | ||
| AK164058.1 | - | 1 - 2 | 138 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164058.1, NM_010605.4 transcripts.
Set Membership: |
|

Kcnj5
Genomic Map