Assay Details
| Gene Symbol: Kcnj5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010605.4 | NP_034735.3 | 3 - 4 | 1375 | 83 | |
| GenBank mRNA | AB019560.1 | - | 1 - 2 | 916 | 83 | |
| AK137600.1 | - | 3 - 4 | 1489 | 83 | ||
| AK164058.1 | - | 3 - 4 | 1376 | 83 | ||
| BC104330.1 | - | 1 - 2 | 934 | 40045462 | 83 | |
| BC104331.1 | - | 1 - 2 | 934 | 40045465 | 83 | |
| BC114976.1 | - | 1 - 2 | 931 | 40045466 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnj5
Genomic Map