Assay Details
| Gene Symbol: Kri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145416.3 | NP_663391.2 | 10 - 11 | 953 | 64 | |
| GenBank mRNA | BC021438.1 | - | 10 - 11 | 943 | 5039543 | 64 |
| CO426749.1 | - | 2 - 3 | 185 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kri1
Genomic Map