Assay Details
| Gene Symbol: Kri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145416.3 | NP_663391.2 | 12 - 13 | 1214 | 78 | |
| GenBank mRNA | BC021438.1 | - | 12 - 13 | 1204 | 5039543 | 78 |
| CO426749.1 | - | 4 - 5 | 446 | 78 | ||
| CX562209.1 | - | 2 - 3 | 241 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kri1
Genomic Map