Assay Details
| Gene Symbol: Kri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145416.3 | NP_663391.2 | 7 - 8 | 624 | 63 | |
| GenBank mRNA | CO426049.1 | - | 4 - 5 | 267 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Kri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145416.3 | NP_663391.2 | 7 - 8 | 624 | 63 | |
| GenBank mRNA | CO426049.1 | - | 4 - 5 | 267 | 63 | |
Set Membership: |
|