Assay Details
| Gene Symbol: Tyk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205312.1 | NP_001192241.1 | 13 - 14 | 2287 | 59 | |
| NM_018793.2 | NP_061263.2 | 13 - 14 | 2218 | 59 | ||
| GenBank mRNA | AK154438.1 | - | 13 - 14 | 2245 | 59 | |
| AK155923.1 | - | 13 - 14 | 2130 | 59 | ||
| AK159303.1 | - | 13 - 14 | 2187 | 59 | ||
| AK167726.1 | - | 13 - 14 | 2197 | 59 | ||
| AK170107.1 | - | 13 - 14 | 2157 | 59 | ||
| AK170940.1 | - | 13 - 14 | 2156 | 59 | ||
| AK171609.1 | - | 13 - 14 | 2130 | 59 | ||
| BC094240.1 | - | 12 - 13 | 2114 | 30610201 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tyk2
Genomic Map