Assay Details
Gene Symbol: Lppr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290299.1 | NP_001277228.1 | 5 - 6 | 726 | 76 | |
NM_144935.1 | NP_659184.1 | 4 - 5 | 698 | 76 | ||
GenBank mRNA | AK082453.1 | - | 5 - 6 | 689 | 76 | |
AY438023.1 | - | 4 - 5 | 698 | 76 | ||
BC018242.1 | - | 4 - 5 | 698 | 4503683 | 76 | |
BC023082.1 | - | 5 - 6 | 703 | 5357160 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
