Assay Details
| Gene Symbol: Icam4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023892.2 | NP_076381.1 | 1 - 2 | 452 | 57 | |
| GenBank mRNA | AF296282.1 | - | 1 - 2 | 371 | 57 | |
| AF296283.1 | - | 1 - 2 | 371 | 57 | ||
| AK019013.1 | - | 1 - 2 | 154 | 57 | ||
| BC116909.1 | - | 1 - 2 | 452 | 40126228 | 57 | |
| BC116913.1 | - | 1 - 2 | 452 | 40126232 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Icam4
Genomic Map