Assay Details
| Gene Symbol: AB124611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198794.1 | NP_001185723.1 | 3 - 4 | 545 | 66 | |
| GenBank mRNA | AB124611.1 | - | 3 - 4 | 445 | 66 | |
| AK137593.1 | - | 4 - 5 | 632 | 66 | ||
| AK154442.1 | - | 3 - 4 | 532 | 66 | ||
| AK162293.1 | - | 3 - 4 | 532 | 66 | ||
| AK168311.1 | - | 3 - 4 | 532 | 66 | ||
| AK170200.1 | - | 3 - 4 | 532 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

AB124611
Genomic Map