Assay Details
| Gene Symbol: AB124611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198794.1 | NP_001185723.1 | 4 - 5 | 568 | 69 | |
| GenBank mRNA | AB124611.1 | - | 4 - 5 | 468 | 69 | |
| AK137593.1 | - | 5 - 6 | 655 | 69 | ||
| AK154442.1 | - | 4 - 5 | 555 | 69 | ||
| AK162293.1 | - | 4 - 5 | 555 | 69 | ||
| AK168311.1 | - | 4 - 5 | 555 | 69 | ||
| AK170200.1 | - | 4 - 5 | 555 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

AB124611
Genomic Map