Assay Details
| Gene Symbol: Spc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026282.5 | NP_080558.1 | 3 - 4 | 412 | 91 | |
| GenBank mRNA | AK011728.1 | - | 3 - 4 | 430 | 91 | |
| AK075950.1 | - | 3 - 4 | 413 | 91 | ||
| AK153185.1 | - | 3 - 4 | 430 | 91 | ||
| BC086683.1 | - | 3 - 4 | 415 | 30676712 | 91 | |
| BC098472.1 | - | 420 | 91 | |||
| BC116238.1 | - | 3 - 4 | 413 | 40091783 | 91 | |
| BC116239.1 | - | 3 - 4 | 412 | 40091784 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spc24
Genomic Map