Assay Details
| Gene Symbol: Raver1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027911.3 | NP_082187.1 | 5 - 6 | 1086 | 81 | |
| GenBank mRNA | AK051444.1 | - | 5 - 6 | 1098 | 82 | |
| AK084478.1 | - | 5 - 6 | 1098 | 81 | ||
| AK138596.1 | - | 5 - 6 | 1098 | 81 | ||
| AK168436.1 | - | 5 - 6 | 1108 | 81 | ||
| BC058271.1 | - | 5 - 6 | 1086 | 6490833 | 81 | |
| BC108392.1 | - | 5 - 6 | 1085 | 6393833 | 81 | |
| Gene Symbol: Gm38431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038081.2 | 5 - 6 | 1106 | 81 | ||
| GenBank mRNA | AK004919.1 | - | 5 - 6 | 1099 | 81 | |
| AY275472.1 | - | 5 - 6 | 1085 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Raver1
Genomic Map