Assay Details
| Gene Symbol: Raver1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027911.3 | NP_082187.1 | 7 - 8 | 1290 | 103 | |
| GenBank mRNA | AK051444.1 | - | 7 - 8 | 1303 | 103 | |
| AK084478.1 | - | 7 - 8 | 1302 | 103 | ||
| AK138596.1 | - | 7 - 8 | 1302 | 103 | ||
| AK168436.1 | - | 7 - 8 | 1312 | 103 | ||
| BC058271.1 | - | 7 - 8 | 1290 | 6490833 | 103 | |
| BC108392.1 | - | 7 - 8 | 1289 | 6393833 | 103 | |
| Gene Symbol: Gm38431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038081.2 | 7 - 8 | 1310 | 103 | ||
| GenBank mRNA | AK004919.1 | - | 7 - 8 | 1303 | 103 | |
| AY275472.1 | - | 7 - 8 | 1289 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Raver1
Genomic Map