Assay Details
| Gene Symbol: Gm38431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038081.2 | 8 - 9 | 1528 | 115 | ||
| GenBank mRNA | AK004919.1 | - | 8 - 9 | 1521 | 115 | |
| AY275472.1 | - | 8 - 9 | 1507 | 115 | ||
| Gene Symbol: Raver1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027911.3 | NP_082187.1 | 8 - 9 | 1508 | 115 | |
| GenBank mRNA | AK051444.1 | - | 8 - 9 | 1521 | 115 | |
| AK084478.1 | - | 8 - 9 | 1520 | 115 | ||
| AK168436.1 | - | 8 - 9 | 1530 | 115 | ||
| AK205393.1 | - | 1 - 2 | 125 | 115 | ||
| BC058271.1 | - | 8 - 9 | 1508 | 6490833 | 115 | |
| BC108392.1 | - | 8 - 9 | 1507 | 6393833 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm38431
Genomic Map