Assay Details
| Gene Symbol: Ilf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042707.2 | NP_001036172.1 | 5 - 6 | 880 | 106 | |
| NM_001042708.2 | NP_001036173.1 | 6 - 7 | 919 | 106 | ||
| NM_001042709.2 | NP_001036174.1 | 5 - 6 | 880 | 106 | ||
| NM_001277321.1 | NP_001264250.1 | 6 - 7 | 919 | 106 | ||
| NM_001277322.1 | NP_001264251.1 | 7 - 8 | 847 | 106 | ||
| NM_010561.3 | NP_034691.2 | 6 - 7 | 919 | 106 | ||
| GenBank mRNA | AB221609.1 | - | 4 - 5 | 595 | 106 | |
| AF098967.1 | - | 7 - 8 | 796 | 106 | ||
| AF497751.2 | - | 6 - 7 | 787 | 106 | ||
| AF497752.2 | - | 5 - 6 | 748 | 106 | ||
| AK048096.1 | - | 5 - 6 | 810 | 106 | ||
| AK077560.1 | - | 6 - 7 | 845 | 106 | ||
| AK128936.1 | - | 6 - 7 | 852 | 106 | ||
| AK150276.1 | - | 6 - 7 | 868 | 106 | ||
| BC047272.1 | - | 6 - 7 | 839 | 6440312 | 106 | |
| BC060599.1 | - | 4 - 5 | 583 | 106 | ||
| DQ104405.1 | - | 6 - 7 | 787 | 106 | ||
| DQ104406.1 | - | 5 - 6 | 748 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ilf3
Genomic Map