Assay Details
| Gene Symbol: Fxyd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111073.2 | NP_001104543.1 | 3 - 4 | 581 | 65 | |
| NM_001287213.1 | NP_001274142.1 | 4 - 5 | 453 | 65 | ||
| NM_008761.4 | NP_032787.1 | 4 - 5 | 441 | 65 | ||
| GenBank mRNA | AI121044.1 | - | 4 - 5 | 441 | 65 | |
| AK003068.1 | - | 4 - 5 | 224 | 65 | ||
| AK155763.1 | - | 3 - 4 | 561 | 65 | ||
| AK169966.1 | - | 4 - 5 | 224 | 65 | ||
| AK170810.1 | - | 4 - 5 | 229 | 65 | ||
| BC013340.1 | - | 4 - 5 | 227 | 65 | ||
| CA528582.1 | - | 4 - 5 | 432 | 65 | ||
| U72680.1 | - | 4 - 5 | 247 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fxyd5
Genomic Map