Assay Details
| Gene Symbol: Tle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083927.1 | NP_001077396.1 | 7 - 8 | 1683 | 66 | |
| NM_001083928.1 | NP_001077397.1 | 7 - 8 | 1653 | 66 | ||
| NM_009389.2 | NP_033415.2 | 7 - 8 | 1653 | 66 | ||
| GenBank mRNA | AK031322.1 | - | 5 - 6 | 460 | 66 | |
| AK122524.1 | - | 7 - 8 | 1161 | 66 | ||
| AK158785.1 | - | 6 - 7 | 973 | 66 | ||
| BC056465.1 | - | 7 - 8 | 664 | 5709252 | 66 | |
| X73360.1 | - | 6 - 7 | 836 | 66 | ||
More Information
Set Membership: |
|

Tle3
Genomic Map