Assay Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018736.2 | NP_061206.1 | 1 - 2 | 47 | 88 | |
| GenBank mRNA | AK041248.1 | - | 1 - 2 | 46 | 88 | |
| AK141214.1 | - | 1 - 2 | 59 | 88 | ||
| AK146234.1 | - | 1 - 2 | 80 | 88 | ||
| AK146370.1 | - | 1 - 2 | 80 | 88 | ||
| AK168180.1 | - | 1 - 2 | 80 | 88 | ||
| BC065144.1 | - | 1 - 2 | 47 | 30627207 | 88 | |
| U58987.1 | - | 1 - 2 | 82 | 88 | ||
| U60318.1 | - | 1 - 2 | 82 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041248.1, AK141214.1, AK146234.1, AK146370.1, AK168180.1, BC065144.1, NM_018736.2, U58987.1, U60318.1 transcripts.
Set Membership: |
|

Mre11a
Genomic Map