Assay Details
| Gene Symbol: Panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019482.2 | NP_062355.2 | 1 - 2 | 627 | 71 | |
| GenBank mRNA | AF207817.1 | - | 1 - 2 | 225 | 71 | |
| AK014033.1 | - | 1 - 2 | 610 | 71 | ||
| AK053871.1 | - | 1 - 2 | 612 | 71 | ||
| AK089764.1 | - | 1 - 2 | 573 | 71 | ||
| AK156194.1 | - | 1 - 2 | 612 | 71 | ||
| BC020413.1 | - | 1 - 2 | 558 | 71 | ||
| BC049074.1 | - | 1 - 2 | 627 | 6809247 | 71 | |
| EU446265.1 | - | 1 - 2 | 627 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Panx1
Genomic Map