Assay Details
| Gene Symbol: Gpr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010287.2 | NP_034417.1 | 1 - 2 | 672 | 77 | |
| GenBank mRNA | AK035356.1 | - | 1 - 2 | 717 | 77 | |
| AK081276.1 | - | 1 - 2 | 685 | 77 | ||
| M80481.1 | - | 1 - 2 | 673 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr83
Genomic Map