Assay Details
| Gene Symbol: Snx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028874.2 | NP_083150.1 | 2 - 3 | 2071 | 106 | |
| GenBank mRNA | AK122231.1 | - | 2 - 3 | 2292 | 106 | |
| BC004635.1 | - | 2 - 3 | 543 | 106 | ||
| BC051164.1 | - | 2 - 3 | 2291 | 106 | ||
| BC063262.1 | - | 2 - 3 | 2071 | 6405897 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Snx19
Genomic Map