Assay Details
| Gene Symbol: Snx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028874.2 | NP_083150.1 | 3 - 4 | 2166 | 62 | |
| GenBank mRNA | AK044307.1 | - | 2 - 3 | 1210 | 62 | |
| AK122231.1 | - | 3 - 4 | 2387 | 62 | ||
| BC004635.1 | - | 3 - 4 | 638 | 62 | ||
| BC051164.1 | - | 3 - 4 | 2386 | 62 | ||
| BC063262.1 | - | 3 - 4 | 2166 | 6405897 | 62 | |
More Information
Set Membership: |
|

Snx19
Genomic Map