Assay Details
| Gene Symbol: Rasgrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174155.1 | NP_001167626.1 | 5 - 6 | 650 | 80 | |
| NM_145149.4 | NP_660131.2 | 5 - 6 | 650 | 80 | ||
| NR_045676.1 | 5 - 6 | 650 | 80 | |||
| GenBank mRNA | AK049562.1 | - | 4 - 5 | 393 | 80 | |
| AK087348.1 | - | 5 - 6 | 602 | 81 | ||
| AK089282.1 | - | 5 - 6 | 597 | 80 | ||
| AK154746.1 | - | 5 - 6 | 640 | 80 | ||
| AK162667.1 | - | 5 - 6 | 651 | 80 | ||
| AK165106.1 | - | 4 - 5 | 388 | 80 | ||
| AY196476.1 | - | 5 - 6 | 510 | 80 | ||
More Information
Set Membership: |
|

Rasgrp4
Genomic Map