Assay Details
| Gene Symbol: Yif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110201.1 | NP_001103671.1 | 1 - 2 | 113 | 82 | |
| GenBank mRNA | AK135529.1 | - | 91 | 82 | ||
| AV482229.1 | - | 1 - 2 | 111 | 82 | ||
| BC109331.2 | - | 1 - 2 | 92 | 40051521 | 82 | |
| BU953116.1 | - | 1 - 2 | 110 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Yif1b
Genomic Map