Assay Details
Gene Symbol: AW551984 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199556.1 | NP_001186485.1 | 2 - 3 | 225 | 61 | |
NM_178737.5 | NP_848852.1 | 2 - 3 | 215 | 61 | ||
GenBank mRNA | AK028282.1 | - | 2 - 3 | 189 | 61 | |
AK049098.1 | - | 2 - 3 | 216 | 61 | ||
AK082321.1 | - | 2 - 3 | 198 | 61 | ||
BC061211.1 | - | 2 - 3 | 127 | 30250574 | 61 | |
CJ161472.1 | - | 2 - 3 | 227 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK028282.1, AK049098.1, AK082321.1, BC061211.1, NM_001199556.1, NM_178737.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
