Assay Details
| Gene Symbol: Tbrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025289.3 | NP_079565.2 | 3 - 4 | 608 | 70 | |
| GenBank mRNA | AK003270.1 | - | 3 - 4 | 536 | 70 | |
| AK010925.1 | - | 3 - 4 | 559 | 70 | ||
| AK131158.1 | - | 3 - 4 | 537 | 70 | ||
| AK131638.1 | - | 2 - 3 | 1037 | 70 | ||
| AK150549.1 | - | 3 - 4 | 535 | 70 | ||
| AK151075.1 | - | 3 - 4 | 486 | 70 | ||
| AK151209.1 | - | 3 - 4 | 502 | 70 | ||
| AK151457.1 | - | 3 - 4 | 520 | 70 | ||
| AK166547.1 | - | 3 - 4 | 530 | 70 | ||
| AK185995.1 | - | 3 - 4 | 327 | 70 | ||
| BC021331.1 | - | 3 - 4 | 619 | 5003215 | 70 | |
| BC040813.1 | - | 3 - 4 | 521 | 4223194 | 70 | |
| BC043022.1 | - | 3 - 4 | 523 | 5688749 | 70 | |
| BC054057.1 | - | 3 - 4 | 518 | 70 | ||
| BC065795.1 | - | 3 - 4 | 521 | 1529903 | 70 | |
| DQ144541.1 | - | 3 - 4 | 440 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbrg1
Genomic Map