Assay Details
| Gene Symbol: Tbrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025289.3 | NP_079565.2 | 5 - 6 | 894 | 76 | |
| GenBank mRNA | AK003270.1 | - | 5 - 6 | 820 | 76 | |
| AK010925.1 | - | 5 - 6 | 844 | 76 | ||
| AK150549.1 | - | 5 - 6 | 821 | 76 | ||
| AK151075.1 | - | 5 - 6 | 772 | 76 | ||
| AK151209.1 | - | 5 - 6 | 788 | 76 | ||
| AK151457.1 | - | 5 - 6 | 806 | 76 | ||
| AK166547.1 | - | 5 - 6 | 816 | 76 | ||
| BC021331.1 | - | 5 - 6 | 905 | 5003215 | 76 | |
| BC040813.1 | - | 5 - 6 | 807 | 4223194 | 76 | |
| BC043022.1 | - | 5 - 6 | 809 | 5688749 | 76 | |
| BC054057.1 | - | 4 - 5 | 1075 | 76 | ||
| BC065795.1 | - | 5 - 6 | 807 | 1529903 | 76 | |
| DQ144541.1 | - | 5 - 6 | 726 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbrg1
Genomic Map