Assay Details
| Gene Symbol: Tbrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025289.3 | NP_079565.2 | 8 - 9 | 1239 | 91 | |
| GenBank mRNA | AK003270.1 | - | 8 - 9 | 1165 | 91 | |
| AK010925.1 | - | 8 - 9 | 1189 | 91 | ||
| AK011268.1 | - | 8 - 9 | 1164 | 91 | ||
| AK131158.1 | - | 5 - 6 | 3449 | 91 | ||
| AK150549.1 | - | 8 - 9 | 1166 | 91 | ||
| AK151075.1 | - | 8 - 9 | 1117 | 91 | ||
| AK151209.1 | - | 8 - 9 | 1133 | 91 | ||
| AK151457.1 | - | 8 - 9 | 1151 | 91 | ||
| AK166547.1 | - | 8 - 9 | 1161 | 91 | ||
| BC021331.1 | - | 8 - 9 | 1250 | 5003215 | 91 | |
| BC040813.1 | - | 8 - 9 | 1152 | 4223194 | 91 | |
| BC043022.1 | - | 8 - 9 | 1154 | 5688749 | 91 | |
| BC054057.1 | - | 7 - 8 | 1420 | 91 | ||
| BC065795.1 | - | 8 - 9 | 1152 | 1529903 | 91 | |
| DQ144541.1 | - | 8 - 9 | 1071 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431944 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431945 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431942 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431941 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021555 | AK033980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021555 | AK131158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021555 | BC043022.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021555 | BC054057.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2021555 | BC040813.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715408 | AK029680.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 397273 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 52498 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117068 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021555 | AK033980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021555 | AK131158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021555 | BC043022.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021555 | BC054057.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2021555 | BC040813.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26431946 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06754123 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06754109 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117068 | - | - |
| Whole Mouse Genome G4122A | A_51_P186531 | DQ144541.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P186531 | BC054057.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P186531 | BC040813.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P186531 | BC065795.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P186531 | AK166547.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101479 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117068 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07622663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07622641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07622642 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07622637 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07622643 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07622648 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431945 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431942 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431944 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26431941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26432001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26431881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08116548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431931 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08116552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07730899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07622655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431945 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08116542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431954 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431895 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07819819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26431927 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10592345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592346 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592348 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592344 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592349 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10592354 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452648_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10592342 | - | - |
| Mouse Genome 430 2.0 Array | 1452648_at | - | - |
| Mouse Genome 430 2.0 Array | 1452648_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21376_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21376_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900017137.mm.1 | NM_025289.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017137.mm.1 | NM_025289.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900017137.mm.1 | TC0900002054.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452648_at | - | - |
| Mouse Expression Set 430 | 1452648_PM_at | - | - |
| Murine Genome U74v2 Set | 98937_PM_at | - | - |
| Murine Genome U74v2 Set | 98937_at | - | - |
| Murine Genome U74v2 Set | 171212_at | - | - |
| Murine Genome U74v2 Set | 170145_at | - | - |
| Murine Genome U74v2 Set | 171212_PM_at | - | - |
| Murine Genome U74v2 Set | 170145_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452648_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452648_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452648_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452648_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 770242 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6520181 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 770242 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006520181 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000770242 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244753 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2667846 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2434144 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2667846 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244753 | - | - |
| mouse-6 v1.1 (Illumina) | scl36022.9.1_13-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0003566.1_12-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 770242 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6520181 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36022.9.1_13-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003566.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6520181 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 770242 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244753 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2667842 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2667842 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244753 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0003566.1_12-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36022.9.1_13-S | - | - |

Tbrg1
Genomic Map