Assay Details
| Gene Symbol: Ttc9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028417.1 | NP_082693.1 | 2 - 3 | 614 | 92 | |
| GenBank mRNA | AK013779.1 | - | 2 - 3 | 617 | 92 | |
| BC048750.1 | - | 2 - 3 | 612 | 92 | ||
| BC089566.1 | - | 2 - 3 | 614 | 6531894 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018405 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018407 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018406 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018373 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358894 | AK013779.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358894 | BC048750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358894 | BC089566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358894 | NM_028417.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358894 | AK013779.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358894 | BC048750.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358894 | BC089566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P358894 | NM_028417.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105323 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150256 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358894 | AK013779.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358894 | BC048750.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358894 | BC089566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358894 | NM_028417.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358894 | AK013779.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358894 | BC048750.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358894 | BC089566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P358894 | NM_028417.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150256 | - | - |
| Whole Mouse Genome G4122A | A_51_P358894 | AK013779.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358894 | BC048750.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358894 | BC089566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358894 | NM_028417.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358894 | AK013779.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358894 | BC048750.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358894 | BC089566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P358894 | NM_028417.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06004665 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06608168 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06004664 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06122741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018411 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018371 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018373 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018407 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018405 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25018406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25018371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018353 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06457756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06091193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06004664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06004673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25018357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06435250 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10551404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10551402 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429416_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10551401 | - | - |
| Mouse Genome 430 2.0 Array | 1429416_at | - | - |
| Mouse Genome 430 2.0 Array | 1429416_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73032_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73032_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700003759.mm.1 | TC0700000466.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700003759.mm.1 | NM_028417.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700003759.mm.1 | NM_028417.1 | Refseq |
| Mouse Expression Set 430 | 1429416_at | - | - |
| Mouse Expression Set 430 | 1429416_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429416_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429416_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 667358 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237237 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1237237 | - | - |

Ttc9b
Genomic Map