Assay Details
| Gene Symbol: Psmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011874.2 | NP_036004.2 | 8 - 9 | 989 | 67 | |
| GenBank mRNA | AK050677.1 | - | 8 - 9 | 967 | 67 | |
| AK077507.1 | - | 8 - 9 | 952 | 67 | ||
| AK153991.1 | - | 8 - 9 | 972 | 67 | ||
| AK160644.1 | - | 8 - 9 | 951 | 67 | ||
| AK160718.1 | - | 8 - 9 | 943 | 67 | ||
| AK167041.1 | - | 8 - 9 | 934 | 67 | ||
| AK167528.1 | - | 8 - 9 | 944 | 67 | ||
| AK169224.1 | - | 8 - 9 | 952 | 67 | ||
| AK169244.1 | - | 8 - 9 | 1452 | 67 | ||
| BC012708.1 | - | 8 - 9 | 950 | 4211677 | 67 | |
| BC092265.1 | - | 8 - 9 | 933 | 3671250 | 67 | |
| BC094063.1 | - | 3 - 4 | 307 | 67 | ||
| L76223.1 | - | 8 - 9 | 940 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc4
Genomic Map