Assay Details
| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033315.2 | NP_001028487.2 | 1 - 2 | 722 | 114 | |
| GenBank mRNA | AB354583.1 | - | 1 - 2 | 466 | 114 | |
More Information
Set Membership: |
|

Hipk4
Genomic Map