| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700003677.mm.1 |
TC0700000462.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003677.mm.1 |
NM_001033315.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003677.mm.1 |
NM_001033315.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003675.mm.1 |
TC0700000462.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003675.mm.1 |
NM_001033315.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003675.mm.1 |
NM_001033315.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003679.mm.1 |
TC0700000462.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003679.mm.1 |
NM_001033315.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003679.mm.1 |
NM_001033315.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003678.mm.1 |
TC0700000462.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003678.mm.1 |
NM_001033315.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003678.mm.1 |
NM_001033315.2 |
Refseq |