Assay Details
| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043317.2 | NP_001036782.1 | 6 - 7 | 1130 | 86 | |
| NM_009845.3 | NP_033975.3 | 4 - 5 | 1005 | 86 | ||
| NR_102723.1 | 6 - 7 | 1144 | 86 | |||
| GenBank mRNA | AF115400.1 | - | 3 - 4 | 774 | 86 | |
| AF115401.1 | - | 3 - 4 | 774 | 86 | ||
| AK143835.1 | - | 6 - 7 | 1068 | 86 | ||
| AK156735.1 | - | 4 - 5 | 1004 | 86 | ||
| AK171589.1 | - | 3 - 4 | 811 | 86 | ||
| AK171990.1 | - | 3 - 4 | 1125 | 86 | ||
| AK172301.1 | - | 4 - 5 | 961 | 86 | ||
| BC046957.1 | - | 6 - 7 | 1056 | 86 | ||
| BC150807.1 | - | 5 - 6 | 1089 | 9087718 | 86 | |
| L02844.1 | - | 5 - 6 | 1276 | 86 | ||
More Information
Set Membership: |
|

Cd22
Genomic Map