Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700024810.mm.1 |
TC0700002779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024810.mm.1 |
NM_001043317.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024810.mm.1 |
NM_001043317.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024810.mm.1 |
NM_009845.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024810.mm.1 |
NM_009845.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024810.mm.1 |
NR_102722.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024810.mm.1 |
NR_102723.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024811.mm.1 |
TC0700002779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024811.mm.1 |
NM_001043317.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024811.mm.1 |
NM_001043317.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024811.mm.1 |
NM_009845.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024811.mm.1 |
NM_009845.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024811.mm.1 |
NR_102722.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024811.mm.1 |
NR_102723.1 |
Refseq |