Assay Details
| Gene Symbol: Cd22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043317.2 | NP_001036782.1 | 10 - 11 | 2195 | 66 | |
| NM_009845.3 | NP_033975.3 | 8 - 9 | 2070 | 66 | ||
| NR_102722.1 | 10 - 11 | 2267 | 66 | |||
| NR_102723.1 | 10 - 11 | 2228 | 66 | |||
| GenBank mRNA | AF102134.1 | - | 7 - 8 | 1839 | 66 | |
| AF115400.1 | - | 7 - 8 | 1839 | 66 | ||
| AF115401.1 | - | 7 - 8 | 1839 | 66 | ||
| AK041230.1 | - | 10 - 11 | 2202 | 66 | ||
| AK143835.1 | - | 10 - 11 | 2133 | 66 | ||
| AK156735.1 | - | 8 - 9 | 2069 | 66 | ||
| AK171589.1 | - | 7 - 8 | 1876 | 66 | ||
| AK172301.1 | - | 8 - 9 | 2026 | 66 | ||
| BC046957.1 | - | 10 - 11 | 2140 | 66 | ||
| BC150807.1 | - | 9 - 10 | 2154 | 9087718 | 66 | |
| L16928.1 | - | 9 - 10 | 2318 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd22
Genomic Map