Assay Details
| Gene Symbol: Dmkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166173.1 | NP_001159645.1 | 17 - 18 | 1672 | 88 | |
| NM_001166174.1 | NP_001159646.1 | 15 - 16 | 1579 | 88 | ||
| NM_028618.2 | NP_082894.1 | 4 - 5 | 263 | 88 | ||
| NM_172899.4 | NP_766487.3 | 16 - 17 | 1624 | 88 | ||
| GenBank mRNA | AK003695.1 | - | 4 - 5 | 266 | 88 | |
| AK081753.1 | - | 17 - 18 | 1674 | 88 | ||
| AY444557.1 | - | 15 - 16 | 1536 | 88 | ||
| AY444558.1 | - | 4 - 5 | 266 | 88 | ||
| AY622962.1 | - | 4 - 5 | 244 | 88 | ||
| AY622963.1 | - | 15 - 16 | 1483 | 88 | ||
| BC064724.1 | - | 16 - 17 | 1617 | 30435974 | 88 | |
More Information
Set Membership: |
|

Dmkn
Genomic Map