Assay Details
| Gene Symbol: Dmkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166173.1 | NP_001159645.1 | 2 - 3 | 726 | 79 | |
| NM_001166174.1 | NP_001159646.1 | 2 - 3 | 726 | 79 | ||
| NM_172899.4 | NP_766487.3 | 2 - 3 | 726 | 79 | ||
| GenBank mRNA | AK081753.1 | - | 2 - 3 | 727 | 80 | |
| AY444557.1 | - | 2 - 3 | 683 | 79 | ||
| AY622963.1 | - | 2 - 3 | 558 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dmkn
Genomic Map