Assay Details
| Gene Symbol: Dmkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166173.1 | NP_001159645.1 | 4 - 5 | 832 | 64 | |
| NM_001166174.1 | NP_001159646.1 | 4 - 5 | 832 | 64 | ||
| NM_172899.4 | NP_766487.3 | 4 - 5 | 832 | 64 | ||
| GenBank mRNA | AK081753.1 | - | 4 - 5 | 834 | 64 | |
| AY444557.1 | - | 4 - 5 | 789 | 64 | ||
| AY622963.1 | - | 4 - 5 | 664 | 64 | ||
| BC064724.1 | - | 4 - 5 | 825 | 30435974 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmkn
Genomic Map