Assay Details
| Gene Symbol: Gapdhs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290631.1 | NP_001277560.1 | 3 - 4 | 458 | 114 | |
| NM_008085.2 | NP_032111.1 | 3 - 4 | 476 | 114 | ||
| GenBank mRNA | M60978.1 | - | 3 - 4 | 458 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gapdhs
Genomic Map