Assay Details
| Gene Symbol: Zbtb32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021397.2 | NP_067372.2 | 4 - 5 | 1082 | 71 | |
| GenBank mRNA | AF241232.1 | - | 3 - 4 | 991 | 71 | |
| BC137687.1 | - | 3 - 4 | 985 | 8860706 | 71 | |
| BC144837.1 | - | 3 - 4 | 747 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb32
Genomic Map