Assay Details
| Gene Symbol: Zbtb32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021397.2 | NP_067372.2 | 5 - 6 | 1244 | 72 | |
| GenBank mRNA | AF241232.1 | - | 4 - 5 | 1153 | 72 | |
| AK015881.1 | - | 3 - 4 | 327 | 72 | ||
| BC137687.1 | - | 4 - 5 | 1147 | 8860706 | 72 | |
| BC144837.1 | - | 4 - 5 | 909 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015881.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytokine production
transcription, DNA-templated
regulation of transcription, DNA-templated
hemopoiesis
T cell proliferation
negative regulation of nucleic acid-templated transcription
nucleic acid binding
DNA binding
transcription corepressor activity
zinc ion binding
metal ion binding
regulation of cytokine production
transcription, DNA-templated
regulation of transcription, DNA-templated
hemopoiesis
T cell proliferation
negative regulation of nucleic acid-templated transcription
nucleic acid binding
DNA binding
transcription corepressor activity
zinc ion binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997509 | NM_021397.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997509 | AF241232.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1997509 | AK015881.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 200300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627036 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166146 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997509 | NM_021397.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997509 | AF241232.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1997509 | AK015881.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P415369 | AF241232.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032451 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032406 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166146 | - | - |
| Whole Mouse Genome G4122A | A_51_P415369 | AF241232.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134916 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166146 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429831 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429842 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429819 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429850 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06429845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06429850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06256953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06401493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06429851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05923679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032491 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05962324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06165539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06327265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06543804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032432 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25032416 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10562045 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562047 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562048 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562049 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10562046 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432459_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10562044 | - | - |
| Mouse Genome 430 2.0 Array | 1432459_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1432459_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58206_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 58206_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700024669.mm.1 | TC0700002769.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024669.mm.1 | NM_021397.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024669.mm.1 | NM_021397.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024668.mm.1 | TC0700002769.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700024668.mm.1 | NM_021397.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700024668.mm.1 | NM_021397.2 | Refseq |
| Affymetrix HT Mouse Genome 430A Array | 1432459_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1432459_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 844045 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5900397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5900397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005900397 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2770119 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2770119 | - | - |
| mouse-6 v1.1 (Illumina) | scl31518.8.1_186-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5900397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31518.8.1_186-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5900397 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2829268 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2829268 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2770119 | - | - |

Zbtb32
Genomic Map