Assay Details
| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172898.3 | NP_766486.1 | 6 - 7 | 1438 | 87 | |
| GenBank mRNA | AK049284.1 | - | 6 - 7 | 1439 | 87 | |
| AK083069.1 | - | 6 - 7 | 1580 | 87 | ||
| AY305303.1 | - | 5 - 6 | 576 | 87 | ||
| BC052773.1 | - | 6 - 7 | 901 | 6314924 | 87 | |
| BC158021.1 | - | 6 - 7 | 946 | 9007327 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kirrel2
Genomic Map