Assay Details
Gene Symbol: Syne4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153577.2 | NP_705805.1 | 4 - 5 | 749 | 82 | |
GenBank mRNA | AA407649.1 | - | 2 - 3 | 220 | 82 | |
BC004761.1 | - | 4 - 5 | 698 | 3155758 | 82 | |
BC023803.1 | - | 4 - 5 | 736 | 5352953 | 82 | |
BC056649.1 | - | 4 - 5 | 688 | 5036575 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
