Assay Details
Gene Symbol: Zfp810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145612.2 | NP_663587.1 | 3 - 4 | 273 | 74 | |
GenBank mRNA | AK032556.1 | - | 3 - 4 | 313 | 74 | |
BC005471.1 | - | 3 - 4 | 273 | 3496918 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK032556.1, BC005471.1, NM_145612.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
