Assay Details
| Gene Symbol: Elavl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010487.2 | NP_034617.1 | 6 - 7 | 1170 | 69 | |
| GenBank mRNA | BC052097.1 | - | 6 - 7 | 1157 | 5695543 | 69 |
| U29148.1 | - | 6 - 7 | 1149 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elavl3
Genomic Map