Assay Details
| Gene Symbol: Lypd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029806.1 | NP_084082.1 | 4 - 5 | 539 | 60 | |
| GenBank mRNA | AK008654.1 | - | 4 - 5 | 542 | 60 | |
| BC107186.2 | - | 4 - 5 | 539 | 40050796 | 60 | |
| BC107187.2 | - | 4 - 5 | 539 | 40050800 | 60 | |
| BC119485.1 | - | 4 - 5 | 539 | 40050803 | 60 | |
| BC119486.1 | - | 4 - 5 | 539 | 40050804 | 60 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2072801 | AK008654.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2072801 | BC107186.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2072801 | BC107187.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2072801 | BC119485.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2072801 | BC119486.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 672735 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120470 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158500 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2072801 | AK008654.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2072801 | BC107186.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2072801 | BC107187.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2072801 | BC119485.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2072801 | BC119486.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158500 | - | - |
| Whole Mouse Genome G4122A | A_51_P199089 | AK008654.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25004443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004417 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06153303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004415 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06215940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06419779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06537506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004433 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06257545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06651839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06306354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06520199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06419776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06527672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06369051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06599643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06520208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004423 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25004430 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10550875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10550873 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453809_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10550870 | - | - |
| Mouse Genome 430 2.0 Array | 1453809_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453809_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76942_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76942_PM_at | - | - |
| Mouse Expression Set 430 | 1453809_at | - | - |
| Mouse Expression Set 430 | 1453809_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453809_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453809_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2798029 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2798029 | - | - |

Lypd5
Genomic Map