Assay Details
Gene Symbol: Zfp61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009561.2 | NP_033587.2 | 2 - 3 | 459 | 59 | |
GenBank mRNA | AK149960.1 | - | 2 - 3 | 254 | 59 | |
BC006618.1 | - | 2 - 3 | 498 | 3967527 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK149960.1, BC006618.1, NM_009561.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
