Assay Details
| Gene Symbol: Chek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007691.5 | NP_031717.2 | 8 - 9 | 924 | 115 | |
| GenBank mRNA | AF016583.1 | - | 7 - 8 | 833 | 115 | |
| AK011258.1 | - | 8 - 9 | 868 | 115 | ||
| AK033179.1 | - | 8 - 9 | 894 | 115 | ||
| AK045336.1 | - | 8 - 9 | 866 | 115 | ||
| AK166675.1 | - | 8 - 9 | 925 | 115 | ||
| BC037613.1 | - | 7 - 8 | 637 | 115 | ||
| BC053394.1 | - | 8 - 9 | 861 | 115 | ||
| BC064445.1 | - | 7 - 8 | 1323 | 115 | ||
| BC100386.1 | - | 7 - 8 | 999 | 30481390 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chek1
Genomic Map