Assay Details
| Gene Symbol: Stt3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008408.4 | NP_032434.3 | 12 - 13 | 1433 | 106 | |
| GenBank mRNA | AK030363.1 | - | 12 - 13 | 1497 | 106 | |
| AK077877.1 | - | 12 - 13 | 1457 | 106 | ||
| AK081547.1 | - | 12 - 13 | 1490 | 106 | ||
| AK129027.1 | - | 12 - 13 | 1471 | 106 | ||
| AK150640.1 | - | 12 - 13 | 1478 | 106 | ||
| AK151541.1 | - | 12 - 13 | 1489 | 106 | ||
| AK151791.1 | - | 12 - 13 | 1489 | 106 | ||
| AK152242.1 | - | 12 - 13 | 1478 | 106 | ||
| AK153840.1 | - | 12 - 13 | 1434 | 106 | ||
| AK153886.1 | - | 12 - 13 | 1534 | 106 | ||
| AK160037.1 | - | 12 - 13 | 1472 | 106 | ||
| AK160607.1 | - | 12 - 13 | 1434 | 106 | ||
| AK168505.1 | - | 12 - 13 | 1479 | 106 | ||
| AK168937.1 | - | 12 - 13 | 1478 | 106 | ||
| AK169669.1 | - | 12 - 13 | 1457 | 106 | ||
| AX799090.1 | - | 12 - 13 | 1476 | 106 | ||
| BC028897.1 | - | 11 - 12 | 1382 | 106 | ||
| BC037612.1 | - | 12 - 13 | 1472 | 5353797 | 106 | |
| BC085313.1 | - | 12 - 13 | 1462 | 30443880 | 106 | |
| L34260.1 | - | 12 - 13 | 1476 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stt3a
Genomic Map