Assay Details
| Gene Symbol: Ei24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199494.1 | NP_001186423.1 | 6 - 7 | 685 | 142 | |
| NM_007915.5 | NP_031941.1 | 6 - 7 | 569 | 142 | ||
| GenBank mRNA | AK145018.1 | - | 6 - 7 | 646 | 142 | |
| AK147776.1 | - | 6 - 7 | 598 | 142 | ||
| AK172233.1 | - | 6 - 7 | 649 | 142 | ||
| AK212828.1 | - | 1 - 2 | 130 | 142 | ||
| BU148129.1 | - | 6 - 7 | 638 | 142 | ||
| U41751.2 | - | 6 - 7 | 573 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ei24
Genomic Map